Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnp
Entrez Gene ID
100753220
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614078: 238847 - 245467 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504637.2 XP_003504685.2 2 - 3 799 96

View Full Product Details