Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Fam83a
Entrez Gene ID
100760967
Gene Name
family with sequence similarity 83, member A
Gene Aliases
I79_025909
Chromosome Location
Chr.Un|NW_003625647: 226 - 705 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003516012.1 XP_003516060.1 - 218 103

View Full Product Details