Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd6
Entrez Gene ID
103161198
Gene Name
BTB (POZ) domain containing 6
Gene Aliases
I79_004723
Chromosome Location
Chr.Un|NW_003613702: 2550703 - 2552954 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007633029.1 XP_007631219.1 3 - 4 313 93

View Full Product Details