Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcbd1
Entrez Gene ID
100767749
Gene Name
pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha
Gene Aliases
I79_006034
Chromosome Location
Chr.Un|NW_003613759: 501779 - 505671 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499899.2 XP_003499947.1 1 - 2 49 82

View Full Product Details