Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Noxo1
Entrez Gene ID
100766881
Gene Name
NADPH oxidase organizer 1
Gene Aliases
I79_007958
Chromosome Location
Chr.Un|NW_003613852: 1595823 - 1600334 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501542.2 XP_003501590.2 3 - 4 1111 81
XM_007642196.1 XP_007640386.1 3 - 4 1138 81
XM_007642198.1 XP_007640388.1 3 - 4 1108 81
XM_007642199.1 XP_007640389.1 4 - 5 400 81
XM_007642200.1 XP_007640390.1 4 - 5 346 81

View Full Product Details