Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcfl5
Entrez Gene ID
100766010
Gene Name
transcription factor-like 5 (basic helix-loop-helix)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613943: 550553 - 575964 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503149.2 XP_003503197.1 3 - 4 392 94

View Full Product Details