Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myoc
Entrez Gene ID
100750409
Gene Name
myocilin, trabecular meshwork inducible glucocorticoid response
Gene Aliases
Myocilin
Chromosome Location
Chr.Un|NW_003614159: 22510 - 30354 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505522.1 XP_003505570.1 2 - 3 724 61

View Full Product Details