Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sssca1
Entrez Gene ID
100771453
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616207: 126013 - 127479 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513905.2 XP_003513953.1 2 - 3 240 114
GenBank mRNA JP057908.1 - 2 - 3 255 114

View Full Product Details