Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrg1
Entrez Gene ID
100762987
Gene Name
proline rich Gla (G-carboxyglutamic acid) 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616749: 52234 - 96772 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514580.2 XP_003514628.1 2 - 3 235 99
GenBank mRNA JI883054.1 - 2 - 3 216 99

View Full Product Details