Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif1b
Entrez Gene ID
100774503
Gene Name
eukaryotic translation initiation factor 1B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614638: 241574 - 242865 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649676.1 XP_007647866.1 2 - 3 395 90
GenBank mRNA JI892363.1 - 2 - 3 395 90

View Full Product Details