Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh1
Entrez Gene ID
100751761
Gene Name
protocadherin 1
Gene Aliases
I79_016060
Chromosome Location
Chr.Un|NW_003614499: 112403 - 123394 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648822.1 XP_007647012.1 1 - 2 836 68

View Full Product Details