Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gprc5a
Entrez Gene ID
100751438
Gene Name
G protein-coupled receptor, class C, group 5, member A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613840: 1671141 - 1696759 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501266.2 XP_003501314.1 1 - 2 608 70
GenBank mRNA JI894765.1 - 1 - 2 306 70

View Full Product Details