Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh12
Entrez Gene ID
100751474
Gene Name
protocadherin 12
Gene Aliases
I79_016057
Chromosome Location
Chr.Un|NW_003614499: 26610 - 45219 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508192.2 XP_003508240.1 3 - 4 3057 89

View Full Product Details