Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pold4
Entrez Gene ID
100761682
Gene Name
polymerase (DNA-directed), delta 4, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614797: 327146 - 328777 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509940.2 XP_003509988.1 2 - 3 395 104
GenBank mRNA JI876200.1 - 2 - 3 404 104
JP052714.1 - 2 - 3 403 104

View Full Product Details