Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim35
Entrez Gene ID
100760599
Gene Name
tripartite motif containing 35
Gene Aliases
I79_012337
Chromosome Location
Chr.Un|NW_003614124: 457299 - 504043 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645702.1 XP_007643892.1 9 - 10 1334 57

View Full Product Details