Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pgam2
Entrez Gene ID
100761944
Gene Name
phosphoglycerate mutase 2 (muscle)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614085: 284624 - 287994 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504748.2 XP_003504796.1 2 - 3 600 63

View Full Product Details