Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcwpw2
Entrez Gene ID
100753274
Gene Name
zinc finger, CW type with PWWP domain 2
Gene Aliases
I79_001626
Chromosome Location
Chr.Un|NW_003613617: 3108857 - 3184095 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648520.1 XP_007646710.1 1 - 2 336 102
XM_007648522.1 XP_007646712.1 1 - 2 336 102

View Full Product Details