Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp26
Entrez Gene ID
100761425
Gene Name
dual specificity phosphatase 26 (putative)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613666: 220789 - 224942 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497842.1 XP_003497890.1 1 - 2 222 83

View Full Product Details