Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nhp2
Entrez Gene ID
100759402
Gene Name
NHP2 ribonucleoprotein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613693: 1971978 - 1976196 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498493.2 XP_003498541.1 2 - 3 319 95
GenBank mRNA JI865332.1 - 2 - 3 303 95
JP044321.1 - 2 - 3 303 95

View Full Product Details