Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nrip3
Entrez Gene ID
100752951
Gene Name
nuclear receptor interacting protein 3
Gene Aliases
I79_018142
Chromosome Location
Chr.Un|NW_003614795: 465473 - 477566 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509910.1 XP_003509958.1 5 - 6 539 74

View Full Product Details