Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klk9
Entrez Gene ID
100752653
Gene Name
kallikrein-related peptidase 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614844: 25237 - 29505 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510191.1 XP_003510239.1 2 - 3 197 83

View Full Product Details