Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adcyap1
Entrez Gene ID
100760233
Gene Name
adenylate cyclase activating polypeptide 1 (pituitary)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614869: 89487 - 93636 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510371.1 XP_003510419.1 2 - 3 249 69

View Full Product Details