Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl22
Entrez Gene ID
100760298
Gene Name
chemokine (C-C motif) ligand 22
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613984: 1384872 - 1388921 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503619.1 XP_003503667.1 2 - 3 198 74

View Full Product Details