Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ten1
Entrez Gene ID
100759443
Gene Name
TEN1 CST complex subunit
Gene Aliases
I79_014343
Chromosome Location
Chr.Un|NW_003614300: 296066 - 308104 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506837.2 XP_003506885.2 3 - 4 312 76

View Full Product Details