The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003516001.2 | XP_003516049.1 | 2 - 3 | 709 | 85 |
GenBank mRNA | JI893739.1 | - | 2 - 3 | 337 | 85 |
JP059100.1 | - | 2 - 3 | 337 | 85 |