Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glrx
Entrez Gene ID
100757688
Gene Name
glutaredoxin (thioltransferase)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613767: 1803819 - 1814087 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500001.2 XP_003500049.1 2 - 3 455 81
GenBank mRNA JI884960.1 - 2 - 3 438 81

View Full Product Details