Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex28
Entrez Gene ID
100772753
Gene Name
testis expressed 28
Gene Aliases
I79_013948
Chromosome Location
Chr.Un|NW_003614255: 57694 - 69623 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506544.1 XP_003506592.1 1 - 2 720 103

View Full Product Details