Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Noto
Entrez Gene ID
100769699
Gene Name
notochord homeobox
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614497: 307522 - 311492 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508177.1 XP_003508225.1 2 - 3 603 61

View Full Product Details