Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rassf3
Entrez Gene ID
100770379
Gene Name
Ras association (RalGDS/AF-6) domain family member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615106: 80852 - 141175 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511411.2 XP_003511459.1 4 - 5 502 79
GenBank mRNA JI884779.1 - 3 - 4 394 79

View Full Product Details