Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnd3
Entrez Gene ID
100755904
Gene Name
Rho family GTPase 3
Gene Aliases
I79_020833
Chromosome Location
Chr.Un|NW_003615373: 83299 - 102015 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512149.2 XP_003512197.1 2 - 3 522 108
GenBank mRNA JP048090.1 - 2 - 3 539 108

View Full Product Details