Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcdhga1
Entrez Gene ID
100757910
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
I79_016073
Chromosome Location
Chr.Un|NW_003614499: 649183 - 656829 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648837.1 XP_007647027.1 2 - 3 317 66

View Full Product Details