Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alx1
Entrez Gene ID
100759154
Gene Name
ALX homeobox 1
Gene Aliases
I79_013911
Chromosome Location
Chr.Un|NW_003614249: 722258 - 743330 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506505.2 XP_003506553.1 3 - 4 655 76
GenBank mRNA JI890876.1 - 2 - 3 263 76
JP063873.1 - 2 - 3 263 76

View Full Product Details