Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ly6g6c
Entrez Gene ID
100762454
Gene Name
lymphocyte antigen 6 complex, locus G6C
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614548: 439057 - 442142 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508480.2 XP_003508528.1 2 - 3 168 61

View Full Product Details