Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100763131
Entrez Gene ID
100763131
Gene Name
chromosome unknown open reading frame, human C16orf80
Gene Aliases
I79_017766
Chromosome Location
Chr.Un|NW_003614730: 49191 - 61930 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509581.2 XP_003509629.1 4 - 5 475 72
GenBank mRNA JP045829.1 - 3 - 4 384 72

View Full Product Details