Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrp14
Entrez Gene ID
100753977
Gene Name
NLR family, pyrin domain containing 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616054: 5562 - 24848 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513642.1 XP_003513690.1 9 - 10 2849 74

View Full Product Details