Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pp2d1
Entrez Gene ID
100774567
Gene Name
protein phosphatase 2C-like domain containing 1
Gene Aliases
I79_006718
Chromosome Location
Chr.Un|NW_003613793: 121879 - 183034 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500463.2 XP_003500511.1 2 - 3 350 88

View Full Product Details