Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lsm6
Entrez Gene ID
100769852
Gene Name
LSM6 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613808: 1700613 - 1712646 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500758.2 XP_003500806.1 2 - 3 166 92
GenBank mRNA JI877246.1 - 1 - 2 97 92
JP046167.1 - 2 - 3 239 92

View Full Product Details