Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnd1
Entrez Gene ID
100767501
Gene Name
Rho family GTPase 1
Gene Aliases
I79_019123
Chromosome Location
Chr.Un|NW_003614949: 308710 - 315714 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510741.2 XP_003510789.1 1 - 2 197 81

View Full Product Details