Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcvrn
Entrez Gene ID
100766076
Gene Name
recoverin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613651: 195034 - 202708 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497491.1 XP_003497539.1 1 - 2 384 66

View Full Product Details