Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrps24
Entrez Gene ID
100761173
Gene Name
mitochondrial ribosomal protein S24
Gene Aliases
I79_011846
Chromosome Location
Chr.Un|NW_003614085: 190649 - 194438 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504745.2 XP_003504793.1 2 - 3 157 84
GenBank mRNA JP058037.1 - 1 - 2 56 84

View Full Product Details