Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf4
Entrez Gene ID
100770179
Gene Name
ring finger protein 4
Gene Aliases
I79_016884
Chromosome Location
Chr.Un|NW_003614595: 389802 - 411134 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508849.2 XP_003508897.1 3 - 4 413 64
GenBank mRNA JI881454.1 - 3 - 4 408 64

View Full Product Details