Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il9r
Entrez Gene ID
100759467
Gene Name
interleukin 9 receptor
Gene Aliases
I79_020464
Chromosome Location
Chr.Un|NW_003615248: 106110 - 116021 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652360.1 XP_007650550.1 5 - 6 631 95

View Full Product Details