Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdf7
Entrez Gene ID
100773313
Gene Name
growth differentiation factor 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613876: 91470 - 203659 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642708.1 XP_007640898.1 2 - 3 533 95

View Full Product Details