Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heca
Entrez Gene ID
100774249
Gene Name
headcase homolog (Drosophila)
Gene Aliases
I79_001968
Chromosome Location
Chr.Un|NW_003613627: 556101 - 575318 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496552.2 XP_003496600.1 3 - 4 1136 92

View Full Product Details