Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sall3
Entrez Gene ID
100765427
Gene Name
spalt-like transcription factor 3
Gene Aliases
I79_008277
Chromosome Location
Chr.Un|NW_003613863: 1188274 - 1200440 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501850.1 XP_003501898.1 2 - 3 114 97

View Full Product Details