Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube2l6
Entrez Gene ID
100756751
Gene Name
ubiquitin-conjugating enzyme E2L 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614216: 511667 - 524959 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506156.2 XP_003506204.1 2 - 3 184 57

View Full Product Details