Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lin28a
Entrez Gene ID
100767698
Gene Name
lin-28 homolog A (C. elegans)
Gene Aliases
I79_017935
Chromosome Location
Chr.Un|NW_003614761: 36832 - 48726 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509731.1 XP_003509779.1 1 - 2 33 97

View Full Product Details