Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Triml2
Entrez Gene ID
100756383
Gene Name
tripartite motif family-like 2
Gene Aliases
I79_021378
Chromosome Location
Chr.Un|NW_003615507: 42220 - 53466 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512585.1 XP_003512633.1 1 - 2 190 69

View Full Product Details