Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkx
Entrez Gene ID
100770324
Gene Name
mohawk homeobox
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613708: 1796138 - 1862192 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498902.2 XP_003498950.1 2 - 3 326 56

View Full Product Details