Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmb3
Entrez Gene ID
100756232
Gene Name
proteasome (prosome, macropain) subunit, beta type, 3
Gene Aliases
I79_005704
Chromosome Location
Chr.Un|NW_003613745: 1532978 - 1540650 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499574.2 XP_003499622.1 2 - 3 325 92
GenBank mRNA JP042495.1 - 2 - 3 272 92

View Full Product Details